Polymyxin B sulphate

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0210056550 0210056501 0210056505 0210056510 0210056525
ICNA0210056550EA 816 EUR
ICNA0210056550 ICNA0210056501 ICNA0210056505 ICNA0210056510 ICNA0210056525
Polymyxin B sulphate
Polymyxin B sulphate

Polymyxin B sulphate is a mixture of Polymyxin B1 and Polymyxin B2 sulphate, obtained from Bacillus polymyxia strains. Polymyxin B inhibits gram-negative organisms in vitro wherein it binds to and interferes with the permeability of the bacterial cytoplasmic membrane and is also reported to inhibit Ca2+ dependent protein kinase C (PKC). Effective concentration is typically 50 mg/l.

Formule: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MDL Nummer: MFCD00131991
CAS nummer: 1405-20-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR