Cytidine free base, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

02101459.5 0210145910 0210145980 0210145905
ICNA02101459.5EA 38.5 EUR
ICNA02101459.5 ICNA0210145910 ICNA0210145980 ICNA0210145905
Cytidine free base, white powder
Cytidine

Soluble in water.

Formule: C₉H₁₃N₃O₅
Smeltpunt: 208…218 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006545
CAS nummer: 65-46-3
EINECS: 200-610-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR