2'-Deoxyadenosine monohydrate ≥98%, powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0210148125 0210148105
ICNA0210148125EA 1027 EUR
ICNA0210148125 ICNA0210148105
2'-Deoxyadenosine monohydrate ≥98%, powder
2'-Deoxyadenosine monohydrate

2′-Deoxyadenosine (2′-dAdo), a deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.


2′-Deoxyadenosine (2′-dAdo), a deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.


Soluble in water (50 mg/ml).

Formule: C₁₀H₁₃N₅O₃·1H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149364
CAS nummer: 16373-93-6
EINECS: 213-488-7

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 186 - 196 °C
UV/Vis: l max (water) 259 ± 2 nm
OD 250/OD 260 0,70 - 0,85
OD 280/OD 260 0,10 - 0,16

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR