DL-Ethionine ∼99%, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0210166201 0210166205
ICNA0210166201EA 84.7 EUR
ICNA0210166201 ICNA0210166205
DL-Ethionine ∼99%, white powder
DL-Ethionine

DL-Ethionine is a racemic mixture of the anti-methylation agent L-ethionine and its enantiomer D-ethionine. DL-Ethionine is used as a dietary supplement to accelerate cholangiocarcinogenesis in vivo. DL-Ethionine is used to induce oxidative stress in liver to study the levels and activities of anti-oxidative enzymes and compounds such as glutathione.


Soluble in 1M hydrochloric acid (50 mg/ml - very slightly hazy colorless solution).


Opgelet: Research Use Only (RUO).

Formule: C₆H₁₃NO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063102
CAS nummer: 67-21-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR