L-Leu-L-Ala, MP Biomedicals

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

02102167.1 0210216701
ICNA02102167.1EA 60.8 EUR
ICNA02102167.1 ICNA0210216701
L-Leu-L-Ala, MP Biomedicals
Eiwitten en peptiden

Soluble in acetic acid 80% (50 mg/ml - clear colourless solution)


  • Synonyms: Leu-Ala; H-Leu-Ala-OH
  • Form: White Crystalline Powder
  • Storage: 0 °C
  • Optical Rotation: [α]20/D +19±3°, c = 2% in methanol


Used as fluorescence intensifier in fluorimetric methods for the determination of phenylalanine.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR