L-Methionine

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

02102291.1 02102291.5 0210229101 02102291.3
ICNA02102291.1EA 71.6 EUR
ICNA02102291.1 ICNA02102291.5 ICNA0210229101 ICNA02102291.3
L-Methionine
L-Methionine
Formule: C₅H₁₁NO₂S
Kookpunt: 278 °C (1013 hPa)
Smeltpunt: 136 °C
Dichtheid: 1,373 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063097
CAS nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR