Methyl-4-hydroxybenzoate, white crystalline powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

02102341.1 02102341.5 0210234101 0210234105
ICNA02102341.1EA 27.9 EUR
ICNA02102341.1 ICNA02102341.5 ICNA0210234101 ICNA0210234105
Methyl-4-hydroxybenzoate, white crystalline powder
Methyl-4-hydroxybenzoate

Methyl p-Hydroxybenzoate is the methyl ester of p-hydroxybenzoic acid. Typically used as a preservative in foods, beverages and cosmetics.


It can be used to inhibit yeast growth and can also be used as an antimicrobial agent for gram-positive anaerobic bacteria. It must be suspended in ethanol first, since it does not dissolve readily in water. It is also indicated to have some pheromonal properties.


Slightly soluble in water (1 g/400 ml); soluble in warm oil (1 g/40 ml) or warm glycerol (1 g/70 ml); freely soluble in alcohol (50 mg/mL methanol - clear, colorless solution), acetone or ether.


Opgelet: Research Use Only (RUO).

Formule: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Kookpunt: 270…280 °C
Smeltpunt: 124…131 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002352
CAS nummer: 99-76-3
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 124 - 131 °C
UV/Vis: l max (methanol) 255 ± 3 nm
Solubility Passes
Acidity Passes
Residue on Ignition ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR