2,4,6-Tris(2-pyridyl)-1,3,5-triazine, dark yellow powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine

02103129.5 0210312925 0210312905 0210312910
ICNA02103129.5EA 38.4 EUR
ICNA02103129.5 ICNA0210312925 ICNA0210312905 ICNA0210312910
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, dark yellow powder
2,4,6-Tris(2-pyridyl)-1,3,5-triazine

Soluble in methanol (100 mg/ml).


2,4,6-Tripyridyl-S-triazine is used for determination of serum iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.


2,4,6-Tris(2-pyridyl)-s-triazine is employed along with ferrous ion to form a complex that allows for spectrophotometric determination of acetaminophen in samples of serum and plasma.


Soluble in methanol (100 mg/ml).

Formule: C₁₈H₁₂N₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006045
CAS nummer: 3682-35-7
EINECS: 222-965-9

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to standard
Melting Point 244 - 250°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR