Monoolein, liquid

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0210488850 0210488880 0210488890 0210488891
ICNA0210488850EA 75.6 EUR
ICNA0210488850 ICNA0210488880 ICNA0210488890 ICNA0210488891
Monoolein, liquid
Monoolein

Glyceryl Monooleate is a surfactant that releases free glycerol and oleic acid upon hydrolysis.


Glyceryl monooleate has been used in liquid crystal studies and research shows that in the presence of monoolein, the penetration of the drug cisplatin is doubled. Monoolein (1-Oleoyl-rac-glycerol) is used in the development of monoolein-based nanoparticulate liquid dispersions as possible vehicles for drug delivery.


Soluble in chloroform (50 mg/ml). Also soluble in hexane.


Opgelet: Research Use Only (RUO).

Formule: C₂₁H₄₀O₄
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00042735
CAS nummer: 111-03-5

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR