Magnesium sulphate, anhydrous ≥98%

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: MgSO4, Magnesium sulphate anhydrous

0215013691 0215013690
ICNA0215013691EA 119 EUR
ICNA0215013691 ICNA0215013690
Magnesium sulphate, anhydrous ≥98%
Magnesium sulphate

Soluble in water.

Formule: MgSO₄
MW: 120.37 g/mol
Smeltpunt: 1127
Dichtheid: 2.65 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011110
CAS nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR