Cefadroxil, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0215058101 0215058105
ICNA0215058101EA 77.5 EUR
ICNA0215058101 ICNA0215058105
Cefadroxil, white powder
Cefadroxil

Antibiotic. Cefadroxil was used to investigate its interaction with hPEPT1 and dipeptide Gly-Sar on basis of steady-state and presteady-state analysis thus testing the hypothesis that variations in drug response among individuals are caused by alterations in the genes involved in drug absorption.


Slightly soluble in water; practically insoluble in alcohol, chloroform, ether.


Opgelet: Research Use Only (RUO). Protect from light.

Formule: C₁₆H₁₇N₃O₅S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00865091
CAS nummer: 50370-12-2

Order Now

Specification Test Results

Identity Test Passes
Potency Report Result (µg/mg)
Melting Point 190 - 205 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR