Guanidiniumchloride ≥99%, white crystalline powder optical density grade

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

0215121025 0215121080 0215121090 0215121091
ICNA0215121025EA 26.3 EUR
ICNA0215121025 ICNA0215121080 ICNA0215121090 ICNA0215121091
Guanidiniumchloride ≥99%, white crystalline powder optical density grade
Guanidiniumchloride

Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins.


This strong denaturant can solubilise insoluble or denatured proteins such as inclusion bodies and be used for the recovery of periplasmic proteins. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. It is also used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties. It is an inhibitor of RNase.

Formule: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smeltpunt: 185
MDL Nummer: MFCD00013026
CAS nummer: 50-01-1
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 180 - 190 °C
OD260 (6,0 M aq soln) ≤0,03
Solubility (6,0 M aq soln) C Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR