1-Heptanesulphonic acid sodium salt, hydrat ~98%, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215123905 0215123925 0215123980
ICNA0215123905EA 40.6 EUR
ICNA0215123905 ICNA0215123925 ICNA0215123980
1-Heptanesulphonic acid sodium salt, hydrat ~98%, white powder
1-Heptanesulphonic acid sodium salt

Sodium 1-heptanesulfonate is a commonly used ion pairing reagent for HPLC, including analyses of peptides and proteins. It has also been used in high performance capillary electrophoresis analysis of peptides.


Soluble in water (10% w/v - clear, colorless solution), ethanol or methanol (0,4% w/v - clear solution).


Opgelet: Research Use Only (RUO).

Formule: H₃C(CH₂)₅CH₂SO₃Na
MDL Nummer: MFCD00007543
CAS nummer: 22767-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
Moisture 7.4 - 9.0%
FTIR Conforms to Standard
Moisture Content 7.4-9.0%
UV/VIS:(60 mg/ml in water adjusted to pH 3)
OD210 nm
<1.0
OD220 nm <0.1
OD254 nm <0.04
OD280 nm <0.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR