2-Mercaptoethanesulphonic acid sodium salt (mesna), white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0215160201 0215160205 0215160210 0215160225
ICNA0215160201EA 49 EUR
ICNA0215160201 ICNA0215160205 ICNA0215160210 ICNA0215160225
2-Mercaptoethanesulphonic acid sodium salt (mesna), white powder
2-Mercaptoethanesulphonic acid sodium salt (mesna)

2-Mercaptoethane sulphonic acid sodium salt is a thermally stable acidic cofactor in the methyl transfer reaction of methanobacterium.


Also acts as a water-soluble disulfide reducing agent.


Soluble in water (50 mg/ml).


Opgelet: Research Use Only (RUO).

Formule: C₂H₅NaO₃S₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007535
CAS nummer: 19767-45-4

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR