1-Octanesulphonic acid sodium salt monohydrate

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate

0215177805 0215177825 0215177880
ICNA0215177805EA 39.1 EUR
ICNA0215177805 ICNA0215177825 ICNA0215177880
1-Octanesulphonic acid sodium salt monohydrate
1-Octanesulphonic acid sodium salt monohydrate
Formule: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007544
CAS nummer: 207596-29-0
EINECS: 226-195-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR