Helix pomatia beta-Glucuronidase, MP Biomedicals

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215228401 0215228402 0215228405 0215228410
ICNA0215228401EA 52.9 EUR
ICNA0215228401 ICNA0215228402 ICNA0215228405 ICNA0215228410
Helix pomatia beta-Glucuronidase, MP Biomedicals
Enzymen

Beta-glucuronidases are members of the glycosidase family of enzymes that catalyse breakdown of complex carbohydrates. Human β-glucuronidase is a type of glucuronidase that catalyses hydrolysis of β-D-glucuronic acid residues from the non-reducing end of mucopolysaccharides, such as heparan sulfate. In human gut β-glucuronidase converts conjugated bilirubin to the unconjugated form for reabsorption.


  • Gene ID: 2990
  • Gene symbol and aliases: GUSB
  • Uniport number: P08236
  • GHS symbol(s): GHS07, GHS08
  • Signal word: danger


β-Glucuronidases are routinely used for the enzymatic hydrolysis of glucuronides from serum plasma and other fluids prior to analysis by enzyme immunoassay, mass spectrometry, gas chromatography, high performance liquid chromatography, or other means. It is used as a reporter gene in GUS assays to monitor gene expression.


β-Glucuronidase from Helix pomatia is a mixture of enzymes. Inhibitors: D-glucuronic acid, D-galacturonic acid, D-glucaro-1, 4-lactone.


Unit definition: Amount that liberates 1,0 µg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5,0 and 37 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR