Microbial Chymostatin, MP Biomedicals

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215284501 0215284505 0215284525 0215284550
ICNA0215284501EA 56.5 EUR
ICNA0215284501 ICNA0215284505 ICNA0215284525 ICNA0215284550
Microbial Chymostatin, MP Biomedicals
Eiwitten en peptiden

Chymostatin ([(S)-1-Carboxy-2-phenylethyl]-carbamoyl-α-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-Gly-X-Phe-al) Source/Species: Microbial A mixture of type A, B, and C. X = Leu (Type A); Ile (Type B); Val (Type C). Ref: 1. Umezawa, H., et al., J. Antibiot., 23: 425 (1970). 2. Tatsuta, K., et al., J. Antibiot., 26: 625 (1973).


Storage temperature: –20 °C

Order Now

Specificaties

Protein/peptide name Chymostatin
Protein synonyms (S)-1-Carboxy-2-phenylethyl]-carbamoyl-a-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-Gly-X-Phe-al
Environmentally Preferable
Species
Bron
CAS No. 9076-44-2
Formulier White to Yellowish Powder
Storage conditions Store at −0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR