Iron(III) chloride, watervrij, black powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

0215349980 0215349991
ICNA0215349980EA 52.3 EUR
ICNA0215349980 ICNA0215349991
Iron(III) chloride, watervrij, black powder
Iron(III) chloride

Ferric chloride is a moderately strong Lewis acid and it is used as a catalyst in organic synthesis.


The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistant properties. Ferric chloride is used in sewage treatment. It is also used as a leaching agent in chloride hydrometallurgy. The ferric chloride test is a traditional colorimetric test for phenols.


Opgelet: Research Use Only (RUO).

Formule: FeCl₃
Kookpunt: ~316 °C (literatuur)
Dichtheid: 2,898 g/cm³ (25 °C) (literatuur)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011005
CAS nummer: 7705-08-0
UN: 1773
ADR: 8,III

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR