4-Bromophenol, crystalline powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0215489083 0215489090 0215489080
ICNA0215489083EA 160 EUR
ICNA0215489083 ICNA0215489090 ICNA0215489080
4-Bromophenol, crystalline powder
4-Bromophenol

Freely soluble in alcohol, chlororform, ether, glacial acetic acid; slightly soluble in water.


Freely soluble in alcohol, chlororform, ether, glacial acetic acid; slightly soluble in water.

Formule: C₆H₅BrO
Kookpunt: 235…236 °C
Smeltpunt: 64…68 °C
Dichtheid: ~ 1,84 g/cm³ (20 °C)
Vlampunt: ≥ 110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002313
CAS nummer: 106-41-2
EINECS: 203-394-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,01413

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR