Cinnarizine, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215498625
ICNA0215498625EA 128 EUR
ICNA0215498625
Cinnarizine, white powder
Cinnarizine

Ca2+ channel blocker; central and peripheral vasodilator


Soluble in chloroform (50 mg/ml).

Formule: C₂₆H₂₈N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00056037
CAS nummer: 298-57-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR