Methyl cellulose, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Cellulose methyl ether

0215549290 0215549280 0215549283 0215549291
ICNA0215549290EA 318 EUR
ICNA0215549290 ICNA0215549280 ICNA0215549283 ICNA0215549291
Methyl cellulose, white powder
Methyl cellulose

Methyl cellulose is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel. It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.


Methyl cellulose is used as a substitute for water-soluble gums,to render paper greaseproof in adhesives,as thickening agent in cosmetics,as protective colloid in emulsions,as binder and stabilizer in foods,as bulk producer in the formulation of dietetic foods,Pharmaceutic aid (suspending agent),Laxative.


Soluble in cold water. Insoluble in hot water.

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081763
CAS nummer: 9004-67-5

Order Now

Specification Test Results

Identity Test Passes
Viscosity (2% aqueous solution at 25 °C) ~15 centipoises
Loss on Drying ≤5,0%
Methoxy groups 27,5 - 31,5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR