New fuchsine C.I. 42520

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0215582380 0215582325
ICNA0215582380EA 91.8 EUR
ICNA0215582380 ICNA0215582325
New fuchsine C.I. 42520
New fuchsine

Soluble in water and ethanol.

Formule: C₂₂H₂₄ClN₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012567
CAS nummer: 3248-91-7
EINECS: 221-831-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR