Propyleencarbonaat, liquid

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215641590 0215641591 0215641594
ICNA0215641590EA 86.6 EUR
ICNA0215641590 ICNA0215641591 ICNA0215641594
Propyleencarbonaat, liquid
Propyleencarbonaat

Soluble in water.


Soluble in water.

Formule: C₄H₆O₃
MDL Nummer: MFCD00005385
CAS nummer: 108-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR