Bromthalein disodium salt, light blue to off white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

0215672625 0215672601 0215672605
ICNA0215672625EA 1075 EUR
ICNA0215672625 ICNA0215672601 ICNA0215672605
Bromthalein disodium salt, light blue to off white powder
Bromthalein disodium salt

Sulfobromophthalein (BSP) is a dye with a high affinity for organic anion transporting polypeptides (OATPs) and has been used as a substrate for multidrug resistance associated protein 2 (Mrp2). BSP is transported into hepatocytes by OATPs and, after conjugation to glutathione, is excreted into bile by Mrp2. It was found to inhibit the aldo-keto reductase ARK1C20.
Sulfobromophthalein (BSP) is used in diagnosis of hepatic disorders. It is also used for the quantitative determination of proteins.


  • Presentation: Light Blue to Off-White Powder
  • Soluble in water (20 mg/mL - clear, colorless solution).
  • Insoluble in ethanol or acetone.
  • Storage temperature: Room Temperature


Sulphobromophthalein (BSP) is used in diagnosis of hepatic disorders. It is also used for the quantitative determination of proteins.

Formule: C₂₀H₈Br₄Na₂O₁₀S₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012442
CAS nummer: 71-67-0

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR