Dexamethasone sodium phosphate ≥97%

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215756594 0215756590 0215756591
ICNA0215756594EA 1712 EUR
ICNA0215756594 ICNA0215756590 ICNA0215756591
Dexamethasone sodium phosphate ≥97%
Dexamethasone sodium phosphate

Soluble in water (200 mg/4 ml - clear, faint yellow solution).


Free Dexamethasone: 1,0%; Phosphate: 1,0%; Water and Alcohol Content: 16,0%


Dexamethasone Sodium Phosphate is also used in the treatment of emetic effects of cancer therapy and epicondylitis.


An anti-inflammatory glucocorticoid, a pro-drug that is converted to dexamethasone in vivo; stimulates glutamine uptake in the cerebral cortex.

Formule: C₂₂H₂₈FNa₂O₈P
MDL Nummer: MFCD00079105
CAS nummer: 2392-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR