EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215798291 0215798283
ICNA0215798291EA 470 EUR
ICNA0215798291 ICNA0215798283
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], powder
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]

Soluble in acetic acid: water (1:1): ≤50 mg/ml, DMF.


Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester) is a homobifunctional, cleavable cross-linking reagent. It is typically, coupled to molecules containing primary amines by amide bonds buffered at pH 7,5 (6,5 - 8,5). The linkage provides a 12-atom spacer.


Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester) can be used to produce succinic acid 2-[3-(doxorubicin-3'-N-yl)-3-oxopropionyloxy]-ethyl ester N-hydroxysuccinimidyl ester with adriamycin at temperature of 20 °C.

Formule: C₁₈H₂₀N₂O₁₂
MDL Nummer: MFCD00049057
CAS nummer: 70539-42-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR