N-Lauroylsarcosine sodium salt ≥94%, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

0219028950 0219028980 0219028990
ICNA0219028950EA 76.2 EUR
ICNA0219028950 ICNA0219028980 ICNA0219028990
N-Lauroylsarcosine sodium salt ≥94%, white powder
N-Lauroylsarcosine sodium salt

N-Lauroylsarcosine is an anionic detergent.


N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoproteins; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungistatic agent in aqueous dispersion (1%).


pH: 7.5 - 8.5 (10% aqueous solution)


Sodium Salt: Soluble in water (100 mg/mL - clear to slightly hazy, colorless solution).

Formule: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042728
CAS nummer: 137-16-6
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥94%
pH (10% aqueous solution) 7,5 - 8,5
Aggregation Number 2
CMC 14,57 mM at 30 °C (Lit.)
Extinction Coefficient EM = 3 (280 nm) (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR