Bleomycin sulphate (mixture) (from Streptomyces verticillus), white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0219030610
ICNA0219030610EA 525 EUR
ICNA0219030610
Bleomycin sulphate (mixture) (from Streptomyces verticillus), white powder
Bleomycin sulphate (mixture)

Bleomycin Sulfate is an atineoplastic antibiotic. A mixture of cytotonic glycopeptide antibiotics which reacts with DNA and causes strand scission; they have also been shown to have a type of oxygen transferase activity. Bleomycin A2 is the main component of the bleomycin employed clinically. Bleomycins differ from one another in the terminal amine and show varying biological activity.


  • Concentration : 10 - 100 µg/mL
  • Key Applications : It is used in research as antibiotics, antifungal
  • Application Areas : Chemicals|Life Sciences
  • Product Type : Biochemicals
  • Biochemical Category : pharmaceuticals
  • Solubility : Very soluble in water, methanol; slightly soluble in ethanol. Practically insoluble in acetone, ethyl acetate, butyl acetate, ether.

Formule: C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00070310
CAS nummer: 9041-93-4

Order Now

Specification Test Results

Identity Test Passes
Source Streptomyces verticillus

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR