Chlortetracycline hydrochloride, 900 u/mg, yellow powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride

0219032705 0219032725 0219032780
ICNA0219032705EA 42.7 EUR
ICNA0219032705 ICNA0219032725 ICNA0219032780
Chlortetracycline hydrochloride, 900 u/mg, yellow powder
Chlortetracycline hydrochloride

Chlortetracycline hydrochloride has antimicrobial action similar to tetracycline hydrochloride, but it is somewhat less active against many Gram-negative organisms. It was first isolated from the culture of Streptomyces aureofacien.


Chlortetracycline is an antibiotic produced by some strains of Streptomyces aureofaciens. Mode of Action: Inhibits protein synthesis (elongation) by preventing binding of aminoacyl-tRNA to the 30S subunit. Antimicrobial spectrum: Gram-positive. Less active against Gram-negative bacterial than tetracycline. Mode of Resistance: Loss of cell wall permeability.


Cell-permeable fluorescent probe for Ca2+ for measuring calcium flux in endoplasmic reticulum and other intracellular stores in situ.


To prepare Stock Solutions: Add the appropriate amount of powder to water while mixing (not the reverse), as adding water to the powders may result in insoluble mixtures that cannot be filtered without losing potency. If desired, the pH may be adjusted, but a basal media with greater than 2 g/L sodium bicarbonate can easily absorb up to 2 % of an acidic antibiotic solution; therefore, an antibiotic solution with an acidic pH would not influence the pH of the media. After ensuring that the resultant solution is transparent, sterile filter and store in small aliquots at appropriate temperature until use. Prior to use of the media, aseptically add the appropriate amount of antibiotic to the basal media in order to obtain the required working concentration. Mix well and sterile filter if asepsis is believed to have been compromised.


The product is tested in 1 M NaOH, giving a clear yellow to brown solution at 50 mg/ml. Its solubility at room temperature in water is reportedly about 8,6 mg/ml, in methanol, 17,4 mg/ml and in ethanol, 1,7 mg/ml. It is soluble in solutions of alkali hydroxides and carbonates. It is practically insoluble in acetone and other organic solvents (Lit.).

Formule: C₂₂H₂₃ClN₂O₈·HCl
MW: 515.34 g/mol
Smeltpunt: 243…247 °C (ontbinding, lit.)
MDL Nummer: MFCD26142699
CAS nummer: 64-72-2

Order Now

Specification Test Results

Identity Test Passes
Specific Activity >900 u/mg
Heavy Metals 100 ppm
Ash ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR