Vincristine sulphate, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

0219068725 0219068705 0219068701
ICNA0219068725EA 1603 EUR
ICNA0219068725 ICNA0219068705 ICNA0219068701
Vincristine sulphate, white powder
Vincristine sulphate

Vincristine Sulfate, a dimeric Vinca alkaloid. Experiments have reported Vincristine Sulfate to demonstrate a capacity to suppress growth of proliferating cells through marked apoptosis. The cell death caused by this agent seems to result in a sustained accumulation of endogenous ceramide levels. Ceramide, has been proposed as a lipid second messenger with specific antiproliferative mediating responses.


Vincristine Sulfate is used to treat acute leukaemia, malignant lymphoma, Hodgkin′s disease, acute erythraemia, acute panmyelosis, breast cancer, Kaposi’s sarcoma and testicular cancer. It is widely used in cancer research. It is used to study multi-drug resistance. It is used to inhibit cell cycle progression at M-phase and to inhibit monoamine oxidase (MAO).


Vincristine is a plant alkaloid that inhibits microtubule assembly by binding tubulin and inducing coiled spiral aggregate formation. It arrests cell cycle in G2/M-phase by blocking mitotic spindle formation. Vincristine triggers Raf-1 activation, phosphorylation of bcl-2-family proteins, induction of p53 expression, and apoptosis in several tumor cell lines. It inhibits VEGF production in leukemia cell lines. It is a substrate for Pgp and CYP3A4. MRP1 transports vincristine in an ATP- and GSH-dependent manner. Vincristine has some immunosuppressant effect. Vincristine may also interfere with amino acid, cyclic AMP, and glutathione metabolism, calmodulin-dependent Ca2+-transport, cellular respiration, and nucleic acid and lipid biosynthesis.


Soluble in water (50% w/v); soluble in methanol; moderately soluble in chloroform; very slightly soluble in ethanol; insoluble in diethyl ether.

Formule: C₄₆H₅₈N₄O₁₄S
MDL Nummer: MFCD00084729
CAS nummer: 2068-78-2
EINECS: 218-190-0
UN: 2811
ADR: 6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR