Pig alpha-Amylase (from Pancreas), MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0219123910 0219123925 0219123950
ICNA0219123910EA 80 EUR
ICNA0219123910 ICNA0219123925 ICNA0219123950
Pig alpha-Amylase (from Pancreas), MP Biomedicals
Enzymen
One unit catalyses the liberation of 1 mmole reducing group (calculated as maltose per minute at pH 6,9 at 20 °C).
  • Enzyme Commission Number: E.C. 3.2.1.1
  • Supplied as a suspension in 0,5 saturated NaCl pH 6,0 containing 3mM CaCl₂