Potassium acetate 99,0, powder_crystalline_white ACS

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid potassium salt

0219142580 0219142591
ICNA0219142580EA 34.7 EUR
ICNA0219142580 ICNA0219142591
Potassium acetate 99,0, powder_crystalline_white ACS
Potassium acetate

Potassium acetate is utilized in the isolation of RNA intermediates and products from in vitro splicing reactions performed with a HeLa cell nuclear extract.Potassium acetate is used as a buffer in molecular biology research, notably in the isolation of DNA.


Solubility: H₂O: 1 M at 20 °C, clear, colorless

Formule: CH₃COOK
MW: 98.14 g/mol
Smeltpunt: 304
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012458
CAS nummer: 127-08-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥99,0%
Heavy Metals <5 ppm
Insoluble Matter <0,005%
pH 6,5 - 9,0 (5% aq soln)
Chloride <0,003%
Phosphate <0,001%
Sulphate <0,002%
Iron <5 ppm
Calcium <0,005%
Magnesium <0,002%
Sodium <0,03%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR