Natriumsulfaat, watervrij ≥99.0%, powder ACS

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0219144405 02191444.5 0219144401
ICNA0219144405EA 191 EUR
ICNA0219144405 ICNA02191444.5 ICNA0219144401
Natriumsulfaat, watervrij ≥99.0%, powder ACS
Natriumsulfaat

Soluble in water (100 mg/ml), yielding a clear, colorless solution.


Chloride (Cl) <0,001%, Nitrogen Compounds (N) <5 ppm, Phosphate (PO4) <0,001%, Calcium (Ca) <0,02%, Magnesium (Mg) <0,005%, Potassium (K) <0,01%,Iron (Fe) <0,001%

Formule: Na₂SO₄
Kookpunt: 1429 °C (1013 hPa)
Smeltpunt: 888 °C
Dichtheid: 2,7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Sodium sulphate (2:1); Disodium sulphate; Natriumsufat; Salt Cake; sulphuric acid, disodium salt; Thenardite; Trona
Form White powder
Storage Store at room temperature (15 to 30 °C)
Assay 99,0%
Heavy Metals 5 ppm (as Pb)
Insoluble Matter 0,01%
Loss on Drying 0,5%
pH 5,2 - 9,2 (5% aq soln)
Refractive Index 1,484 (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR