Calcium hydrogen orthophosphate, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic

0219380480 0219380490 0219380491
ICNA0219380480EA 28.2 EUR
ICNA0219380480 ICNA0219380490 ICNA0219380491
Calcium hydrogen orthophosphate, white powder
Calcium hydrogen orthophosphate

Calcium phosphate is used in animal feeds, as a mineral supplement, as an antacid, in the manufacturing of glass, in dental products and in fertilizers. It can be used as a dietary supplement, and as an antacid.


Opgelet: Research Use Only (RUO).

Formule: CaHPO₄
MW: 136.06 g/mol
Smeltpunt: ∼370 °C (ontbinding, lit.)
Dichtheid: ∼2.3 g/cm³ (20 °C) (literatuur)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010909
CAS nummer: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR