Acrylamide ≥99%, white crystalline powder for molecular biology

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

0219398280 0219398290 0219398291
ICNA0219398280EA 83.5 EUR
ICNA0219398280 ICNA0219398290 ICNA0219398291
Acrylamide ≥99%, white crystalline powder for molecular biology
Acrylamide

Storage: Room Temperature, desiccate.
Acrylamide is prepared on an industrial scale by the hydrolysis of acrylonitrile by nitrile hydratase.
Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers. It polymerizes in the presence of free radicals in aqueous solutions. It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.
Acrylamide is an extremely hazardous substance in either the powder form or liquid form. Once polymerized, the polyacrylamide is considered safe to handle under normal laboratory conditions. Acrylic Acid Content -≤0.001%


  • Molecular Biology Reagent Grade
  • Presentation: White Crystalline Powder
  • Auto Ignition: 425 °C/795 °F (lit.)
  • Vapor Density: 2.5 (air = 1) (lit.)
  • Vapor Pressure: ca. 20 hPa at 20 °C (lit.)
  • Refractive Index: 1.460 (lit.)
  • UV/Visivle Absorbance: OD290 (1% aq soln) ≤0.10

Formule: C₃H₅NO
Kookpunt: 125 °C (25 mmHg)) (literatuur)
Smeltpunt: 82…92 °C
Dichtheid: 1,127 g/cm³ (25 °C) (literatuur)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008032
CAS nummer: 79-06-1
UN: 2074
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
OD290 (1% aq soln) ≤0.10
Acrylic Acid Content ≤0.001%
pH (1% aq soln) 5.0-7.0
Solubility (1% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR