Dithiothreitol (DTT, Cleland's reagent) ≥99% for molecular biology

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

02194821.2 0219482150 0219482101 0219482105 0219482110 0219482125
ICNA02194821.2EA 34 EUR
ICNA02194821.2 ICNA0219482150 ICNA0219482101 ICNA0219482105 ICNA0219482110 ICNA0219482125
Dithiothreitol (DTT, Cleland's reagent) ≥99% for molecular biology
Dithiothreitol (DTT, Cleland's reagent)

Storage temperature: +4°C


pH: (0.1 M in H2O, 25°C): 4.0 to 6.0 (Lit.)

Formule: C₄H₁₀O₂S₂
Kookpunt: ∼125 °C (1013 hPa)
Smeltpunt: 42…44 °C
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004877
CAS nummer: 3483-12-3
EINECS: 222-468-7
UN: 3335
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR