Glycine ≥99% for molecular biology

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0219482580 0219482590 0219482591 0219482594
ICNA0219482580EA 35.2 EUR
ICNA0219482580 ICNA0219482590 ICNA0219482591 ICNA0219482594
Glycine ≥99% for molecular biology
Glycine

Storage: Store at Room Temperature (15-30 °C)
Glycine is a non-essential amino acid. It is only amino acid with no asymmetric carbon and thus is not chiral. It is the major inhibitory neurotransmitter. It is involved in the biosynthesis of the porphyrin rings of hemes and chlorophylls.
Glycine is commonly used in buffer solutions, in electrophoresis and preparative chromatography. A study of the folding of monoclonal antibodies in the presence of glycine and their subsequent purification has been published. The use of glycine in the purification of lipopolysaccharides, lipooligosaccharides, and lipid A has been reported. It is an amino acid for use in cell culture media development applications and existing media formulations. Glycine is commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots, a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments. The growth requirements of various microorganisms is reported in the Handbook of Microbiology.
Glycine is a non-chiral amino acid that can be synthesized in the body from the amino acid serine by Serine Hydroxymethyltransferase. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.


  • Molecular Biology Reagent Grade
  • Presentation: Powder
  • pKa: 2.35 at 25 °C (Lit.)
  • Soluble in water (250 mg/mL at 25 °C; 672 mg/mL at 100 °C); slightly soluble in alcohol and ethanol (95%); almost insoluble in ether.

Formule: NH₂CH₂COOH
Dichtheid: 1,16 g/mL (literatuur)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008131
CAS nummer: 56-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Loss on Drying ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR