Vancomycin hydrochloride (from Streptomyces orientalis)

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

02195540.1 02195540.2 0219554001 0219554005
ICNA02195540.1EA 69.6 EUR
ICNA02195540.1 ICNA02195540.2 ICNA0219554001 ICNA0219554005
Vancomycin hydrochloride (from Streptomyces orientalis)
Vancomycin hydrochloride

Vancomycin hydrochloride inhibits bacterial mucopeptide biosynthesis.


  • Potency approx. 1000 µg/mg
  • pH 2,5 to 4,5 (5% aqueous solution)
  • Storage temperature +4 °C
  • pH: 2.5 - 4.5 (5% aq solution)

Formule: C₆₆H₇₅Cl₂N₉O₂₄·HCl
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03613611
CAS nummer: 123409-00-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR