Coconut oil, semi-solid, hydrogenated

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0290140401 0290140405
ICNA0290140401EA 59.1 EUR
ICNA0290140401 ICNA0290140405
Coconut oil, semi-solid, hydrogenated
Coconut oil

Coconut oil is a common ingredient of animal diet.


  • Free fatty acids (as Oleic): 0,05% max.


It is considered an edible oil and has several purposes in food, industry, and medicine.

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00130817
CAS nummer: 8001-31-8

Order Now

Specification Test Results

Identity Test Passes

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR