Zetmeel (from corn), fine white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0290295625 0290295680
ICNA0290295625EA 89.7 EUR
ICNA0290295625 ICNA0290295680
Zetmeel (from corn), fine white powder
Zetmeel

Corn starch is a component used in formulating animal research diets.

Formule: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082026
CAS nummer: 9005-25-8

Order Now

Specification Test Results

Identity test Passes
Loss on Drying ≤15%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR