Brewers yeast, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0290331205 0290331225 0290331280
ICNA0290331205EA 84.7 EUR
ICNA0290331205 ICNA0290331225 ICNA0290331280
Brewers yeast, MP Biomedicals
Celcultuursupplementen en -additieven
Brewers yeast is often used as a source of B-complex vitamins, chromium, and selenium. It is also can be used to bake bread, ferment wine, beer and ethanol production.