Fenol vloeibaar ≥99.5%, clear, colorless solution, Aquaphenol

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11AQUAPH01 11AQUAPH02 11AQUAPH03
ICNA11AQUAPH01EA 221 EUR
ICNA11AQUAPH01 ICNA11AQUAPH02 ICNA11AQUAPH03
Fenol vloeibaar ≥99.5%, clear, colorless solution, Aquaphenol
Fenol vloeibaar

Aquaphenol™ is a solution of double-distilled phenol saturated with ultra-pure water (approx 30%). It does not contain antioxidant or additives. Aquaphenol™ has an acid pH and can be used directly for RNA extraction. For DNA extraction, pH can be quickly adjusted to 7,5 or 8 using pH maker solutions and protocols included.


  • Concentration: Phenol (double distilled) saturated with ultra pure water (30% water v/v)
  • Partition Coefficient: 1.47 (Lit.)
  • Presentation: Clear, Colorless Solution
  • pH: 4.5 - 5 (Lit.)
  • Store at +4 °C. Store under nitrogen. Protect from Light.

Formule: C₆H₅OH
MW: 94.11 g/mol
Dichtheid: 1.03 g/cm³ (20 °C) (literatuur)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002143
CAS nummer: 108-95-2
UN: 2821
ADR: 6.1 III

Order Now

Specification Test Results

Iron ≤0.0001%
Heavy Metals ≤0.0005%
Water Content (KF) ~30%
RNase, DNase, Protease Not Detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR