Sieves for Pilotina Mills
Leverancier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
U106469 U106470 U106471
IKAAU106469EA 0 EUR
IKAAU106469 IKAAU106470 IKAAU106471
Sieves for Pilotina Mills
Zeven Zeven voor molens
Interchangeable sieves for insertion into the grinding heads ensure maximum particle size filtering.
All sieves are made of stainless steel 1.4301.