Anti-YAP1 Mouse Monoclonal Antibody [clone: 63.7]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ILM3920-C01 ILM3920-C05 ILM3920-C1
ILM3920-C01EA 186 EUR
ILM3920-C01 ILM3920-C05 ILM3920-C1
Anti-YAP1 Mouse Monoclonal Antibody [clone: 63.7]
Antilichamen
Type: Primary
Antigen: YAP1
Clonality: Monoclonal
Clone: 63.7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR