Anti-CDH1 Mouse Monoclonal Antibody [clone: MX020]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ILM7002-C01 ILM7002-C05 ILM7002-C1
ILM7002-C01EA 211 EUR
ILM7002-C01 ILM7002-C05 ILM7002-C1
Anti-CDH1 Mouse Monoclonal Antibody [clone: MX020]
Antilichamen
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: MX020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR