Manual Vacuum Regulation Turret, Flush Mount
Leverancier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
700562
ILMV700562EA 267 EUR
ILMV700562
Manual Vacuum Regulation Turret, Flush Mount
Ventielen Vacuümventielen
Open/close the vacuum line and easily control flow rate.
  • Dimensions: 690 mm
  • Diameter: 820 mm protrusion from wall and 20 mm into wall
Turret has stepped hose barb that accepts 6,35 - 9,52 mm vacuum hose (DN 8-10).