tempmate® i1 Single Use Indicator, 1 to 40 °C no start delay, no alarm delay
Leverancier: tempmate
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
I1CS400
IMECI1CS400EA 340 EUR
IMECI1CS400
tempmate® i1 Single Use Indicator, 1 to 40 °C no start delay, no alarm delay
Dataloggers
Single use temperature indicator for reliable low-cost cold chain monitoring.
  • Product-related temperature monitoring along
  • the entire cold chain
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • No additional hardware or software required
  • Storage time four years
  • Intelligent LEDs for status monitoring
tempmate.®-i1 indicators are specially designed for temperature monitoring in pharmaceutical cold chain logistics. As preprogrammed device tempmate.®-i1 can be instantly used to control your temperature sensitiveMore Product Information