Anti-IgG Goat Polyclonal Antibody (DyLight® 594)

Leverancier: ImmunoReagents
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IR1672
IMMRIR1672EA 87.5 EUR
IMMRIR1672
Anti-IgG Goat Polyclonal Antibody (DyLight® 594)
Antilichamen
The ImmunoReagents DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-color applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593 nm / 618 nm Emission Color = Red (Similar Dyes: Alexa Fluor 594, Texas Red).

  • Origin: US

ImmunoReagents antibodies are suitable forMore Product Information
Order Now

Specificaties

antigeen symbool IgG
Type antilichaam Ondergeschikt,Immunoglobulin G
Environmentally Preferable
Type antilichaam Ondergeschikt,Immunoglobulin G
Clonality Polyklonaal
Conjugatie DyLight® 594
Reactivity Human
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunoNeerslag Yes
ImmunoFluorescentie Yes
Flowcytometrie Yes
Immunogeen Human IgG
Formaat Purified IgG
Formulier Lyophilized
Purification Affinity purified using solid phaseHuman IgG
Concentratie 1,0 mg/ml (E 1% at 280 nm = 13,0)
Kruisadsorptie No
Opslagbuffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
Opslagtemperatuur 2…8 °C
Shipping temperature 2…8 °C
Tested applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR