Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverancier: ImmunoReagents
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
IR2340
IMMRIR2340EA 79.3 EUR
IMMRIR2340
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information