Filter Fleece
Leverancier: PIEPER FILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PPV75-37-100
INFIPPV75-37-100EA 410 EUR
INFIPPV75-37-100
Filter Fleece
Filters Membraanfilters
Filter fleece is suitable for drainage under membranes. It is also used in chipping processes to keep the coolant free of the chips. The fleece is also suitable for coarse air filtration.
  • Polypropylene fleece
  • Thickness: 75 µm
  • Sampling according to IFA 7512, IFA 6172